missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanecarbonitrile, 98%

Product Code. 11499013
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499013 250 g 250g
11479013 10 g 10g
11489013 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499013 Supplier Thermo Scientific Alfa Aesar Supplier No. A11125.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5500-21-0 | C4H5N | 67.091 g/mol

Cyclopropanecarbonitrile is used to prepare cis and trans crotonitrile and allyl cyanide. Further, it reacts with phenylmagnesium bromide to give cyclopropyl-phenyl ketone-imine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropanecarbonitrile is used to prepare cis and trans crotonitrile and allyl cyanide. Further, it reacts with phenylmagnesium bromide to give cyclopropyl-phenyl ketone-imine.

Solubility
Miscible with water.

Notes
Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5500-21-0
Molecular Formula C4H5N
Molecular Weight (g/mol) 67.091
MDL Number MFCD00001269
InChI Key AUQDITHEDVOTCU-UHFFFAOYSA-N
Synonym cyclopropyl cyanide, cyanocyclopropane, cyclopropylnitrile, cyclopropanenitrile, cyclopropane cyanide, 1-cyanocyclopropane, cyclopropane carbonitrile, cyclopropylcyanide, 1-cyanocyclopropyl, cyclopropancarbonitril
PubChem CID 79637
IUPAC Name cyclopropanecarbonitrile
SMILES C1CC1C#N

Specifications

Density 0.905
Boiling Point 134°C to 135°C
Flash Point 32°C (89°F)
Odor Recognizable
Refractive Index 1.421
Quantity 250 g
UN Number UN3275
Beilstein 635790
Solubility Information Miscible with water.
Formula Weight 67.09
Percent Purity 98%
Chemical Name or Material Cyclopropanecarbonitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.