missing translation for 'onlineSavingsMsg'
Learn More

Cyclophosphamide Monohydrate, LoGiCal

Product Code. 30462114
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30462114

Brand: LoGiCal LGCFOR0822.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclophosphamide Monohydrate
CAS 6055-19-2
Analyte or Component Names Cyclophosphamide Monohydrate
Shipping Condition Room Temperature
Molecular Formula C7 H15 Cl2 N2 O2 P . H2 O
Certifications/Compliance ISO 9001
Synonym Cyclophosphamide monohydrate, 2H-1,3,2-Oxazaphosphorin-2-amine, N,N-bis(2-chloroethyl)tetrahydro-, 2-oxide, monohydrate, 2H-1,3,2-Oxazaphosphorine, 2-[bis(2-chloroethyl)amino]tetrahydro-, 2-oxide, monohydrate, 2-[Bis(2-chloroethyl)amino]tetrahydro-2H-1,3,2-oxazaphosphorine-2-oxide monohydrate, Cyclophosphamide hydrate, Cyclophosphamide Monohydrate, Endoxan monohydrate
InChI Formula InChI=1S/C7H15Cl2N2O2P.H2O/c8-2-5-11(6-3-9)14(12)10-4-1-7-13-14;/h1-7H2,(H,10,12);1H2
Solution Type Neat
SMILES O.ClCCN(CCCl)P1(=O)NCCCO1
IUPAC Name N,N-bis(2-chloroethyl)-2-oxo-1,3,2λ^{5}-oxazaphosphinan-2-amine;hydrate
Industry Type Clinical Testing
Molecular Weight (g/mol) 279.1
Quantity 10 mg
Formula Weight 278.0354 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.