missing translation for 'onlineSavingsMsg'
Learn More

Cyclophosphamide EP Impurity F (Cyclophosphamide USP Related Compound C), Mikromol™

Product Code. 30051626
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051626 100 mg 100mg
30052360 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30051626 Supplier Mikromol Supplier No. MM0822.03
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Cyclophosphamide EP Impurity F (Cyclophosphamide USP Related Compound C)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-aminopropyl dihydrogen phosphate
CAS 1071-28-9
Analyte or Component Names 3-aminopropyl dihydrogen phosphate
Shipping Condition Room Temperature
Molecular Formula C3 H10 N O4 P
Certifications/Compliance ISO 9001
Synonym 3-Aminopropyl Dihydrogen Phosphate, 1-Propanol, 3-amino-, 1-(dihydrogen phosphate), 1-Propanol, 3-amino-, dihydrogen phosphate (ester) (6CI,7CI,8CI,9CI), 3-Aminopropyl dihydrogen phosphate, 3-Aminopropyl monophosphate, Cyclophosphamide Related Compound C
InChI Formula InChI=1S/C3H10NO4P/c4-2-1-3-8-9(5,6)7/h1-4H2,(H2,5,6,7)
SMILES NCCCOP(=O)(O)O
IUPAC Name 3-aminopropyl dihydrogen phosphate
Molecular Weight (g/mol) 155.09
Quantity 100 mg
Formula Weight 155.0347 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.