missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cyclophilin F Monoclonal Antibody (E11AE12BD4)
GREENER_CHOICE

Product Code. 10458653
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10458653 100 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10458653 Supplier Invitrogen™ Supplier No. 455900

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Mouse Monoclonal Antibody

Positive controls: Isolated mitochondria from Human heart, Bovine heart, Rat heart, Mouse heart, HepG2 cells; Cultured Human embryonic lung-derived fibroblasts (strain MRC5); Human cerebellum tissue; HL60 cells.

The protein encoded by this gene is a member of the peptidyl-prolyl cis-trans isomerase (PPIase) family. PPIases catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and accelerate the folding of proteins. This protein is part of the mitochondrial permeability transition pore in the inner mitochondrial membrane. Activation of this pore is thought to be involved in the induction of apoptotic and necrotic cell death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclophilin F
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone E11AE12BD4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation HEPES buffered saline with 0.02% sodium azide
Gene Ppif
Gene Accession No. P29117, P30404, P30405, Q99KR7
Gene Alias AW457192; cyclophilin 3; cyclophilin D; Cyclophilin F; CYP3; CypD; CyP-D; CyP-F; CyP-M; mitochondrial cyclophilin; mitochondrial Cyclophilin D; mitochondrial peptidyl-prolyl cis-trans isomerase; Peptidyl-prolyl cis-trans isomerase F; peptidyl-prolyl cis-trans isomerase F, mitochondrial; peptidyl-prolyl cis-trans isomerase, mitochondrial; peptidylprolyl isomerase F; peptidylprolyl isomerase F (cyclophilin F); PPIase; PPIase F; PPIF; Rotamase F
Gene Symbols Ppif
Host Species Mouse
Immunogen Recombinant rat Cyclophilin F.
Purification Method Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10105, 105675, 282819, 414346
Target Species Human, Mouse, Rat, Bovine
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.