missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclophilin-F Antibody (1F5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-28608
This item is not returnable.
View return policy
Description
Cyclophilin-F Monoclonal specifically detects Cyclophilin-F in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Cyclophilin-F | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50-1:100 | |
| Cyclophilin F, CYP3FLJ90798, Cyp-D, hCyP3, MGC117207, mitochondrial, peptidyl-prolyl cis-trans isomerase, mitochondrial, peptidylprolyl isomerase F, peptidylprolyl isomerase F (cyclophilin F) | |
| Mouse | |
| 22 kDa | |
| 0.1 mL | |
| Proteases & Other Enzymes | |
| 10105 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1F5 | |
| Unconjugated | |
| P30405 | |
| PPIF | |
| Recombinant human PPIF (30-207aa) purified from E. coli (NP_005720). | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction