missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cyclophilin C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106403
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CYPC.
PPIC is a member of the peptidyl-prolyl cis-trans isomerase (PPIase) family. PPIases catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and accelerate the folding of proteins. Similar to other PPIases, this protein can bind immunosuppressant cyclosporin A.
Specifications
| Cyclophilin C | |
| Polyclonal | |
| Unconjugated | |
| Ppic | |
| Cyclophilin C; CyP-20c; CYPC; parvulin; peptidyl-prolyl cis-trans isomerase C; peptidylprolyl isomerase C; peptidylprolyl isomerase C (cyclophilin C); PPIase C; Ppic; Rotamase C | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 19038, 291463, 5480 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P30412, P45877 | |
| Ppic | |
| A synthesized peptide derived from human PPIC(Accession P45877), corresponding to amino acid residues V90-D140. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction