missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cyclophilin C Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569300
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%.
PPIC is a member of the peptidyl-prolyl cis-trans isomerase (PPIase) family. PPIases catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and accelerate the folding of proteins. Similar to other PPIases, this protein can bind immunosuppressant cyclosporin A.
Specifications
| Cyclophilin C | |
| Polyclonal | |
| Unconjugated | |
| Ppic | |
| Cyclophilin C; CyP-20c; CYPC; parvulin; peptidyl-prolyl cis-trans isomerase C; peptidylprolyl isomerase C; peptidylprolyl isomerase C (cyclophilin C); PPIase C; Ppic; Rotamase C | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 19038 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P30412 | |
| Ppic | |
| synthetic peptide directed towards the C-terminal region of Cyclophilin C. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction