missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclophilin C Antibody (3C6), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-97946CL1
This item is not returnable.
View return policy
Description
Cyclophilin C Monoclonal antibody specifically detects Cyclophilin C in Human samples. It is validated for Flow Cytometry,ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| Cyclophilin C | |
| Monoclonal | |
| CoraFluor 1 | |
| Cyclophilin C, CYPCMGC3673, EC 5.2.1.8, parvulin, peptidyl-prolyl cis-trans isomerase C, peptidylprolyl isomerase C (cyclophilin C), PPIase C, Rotamase C | |
| Recombinant human Cyclophilin C (39-197aa) protein purified from E. coli | |
| 0.1 mL | |
| Immunology, Signal Transduction | |
| 5480 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b κ |
| Flow Cytometry, ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 3C6 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction