missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cyclophilin B Recombinant Rabbit Monoclonal Antibody (K01_1Q06)
GREENER_CHOICE

Product Code. 30830131
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830131 100 μL
30828056 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830131 Supplier Invitrogen™ Supplier No. MA563726

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: KVESTKTDSR DKPLK.

Immunophilins are a family of soluble cytosolic receptors capable of binding to one of two major immunosuppressant agents - cyclosporin A (CsA) or FK506. Proteins which bind FK506 are termed FK506 Binding Proteins (FKBP's) and those which bind cyclosporin A are called cyclophilins (CyP). Both CyP:CsA and FKBP:FK506 complexes have been shown to inhibit calcineurin, a calcium and calmodulin dependent protein phosphatase which has been implicated as an important signaling enzyme in T-cell activation. Thus, providing a possible mechanism of immunosuppression by CsA and FK506. Immunophilins function as peptidyl prolyl cis-trans-isomerases (PPIase) whose activity is inhibited by their respective immunosuppressant compounds. As PPIase's, immunophilins accelerate folding of some proteins both in vivo and in vitro by catalyzing slow steps in the initial folding and rearrangement of proline containing proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclophilin B
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1Q06
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene PPIB
Gene Accession No. P23284, P24368, P24369
Gene Alias AA408962; AA553318; AI844835; Cphn2; Cphn-2; cyclophilin B; cyclophilin-like protein; Cyp B; CyP-20b; CYPB; CYP-S1; EC 5.2.1.8; epididymis secretory protein Li 39; HEL-S-39; OI9; Peptidyl-prolyl cis-trans isomerase B; peptidylprolyl isomerase B; peptidylprolyl isomerase B (cyclophilin B); PPIase; PPIase B; Ppib; Rotamase; Rotamase B; S-cyclophilin; S-cyclophylin; SCYLP
Gene Symbols PPIB
Host Species Rabbit
Immunogen A synthetic peptide of human Cyclophilin B (aa 181-195).
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19035, 5479, 64367
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.