Learn More
Invitrogen™ Cyclophilin B Polyclonal Antibody, Alexa Fluor™ 488
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA1027AA488
Description
PA1027AA488 detects cyclophilin B (CyPB) in mouse, rat, canine, and human tissues. The PA1027AA488 immunogen is a synthetic peptide corresponding to residues C(194) G K I E V E K P F A I A K E(208) of human CyPB. This sequence is almost completely conserved between human, mouse, rat, bovine, and chicken. PA1027AA488 immunizing peptide (Cat. # PEP-058) is available for use in neutralization and control experiments.
Immunophilins are a family of soluble cytosolic receptors capable of binding to one of two major immunosuppressant agents - cyclosporin A (CsA) or FK506. Proteins which bind FK506 are termed FK506 Binding Proteins (FKBP's) and those which bind cyclosporin A are called cyclophilins (CyP). Both CyP:CsA and FKBP:FK506 complexes have been shown to inhibit calcineurin, a calcium and calmodulin dependent protein phosphatase which has been implicated as an important signaling enzyme in T-cell activation. Thus, providing a possible mechanism of immunosuppression by CsA and FK506. Immunophilins function as peptidyl prolyl cis-trans-isomerases (PPIase) whose activity is inhibited by their respective immunosuppressant compounds. As PPIase's, immunophilins accelerate folding of some proteins both in vivo and in vitro by catalyzing slow steps in the initial folding and rearrangement of proline containing proteins.
Specifications
| Cyclophilin B | |
| Polyclonal | |
| Alexa Fluor 488 | |
| PPIB | |
| AA408962; AA553318; AI844835; Cphn2; Cphn-2; cyclophilin B; cyclophilin-like protein; Cyp B; CyP-20b; CYPB; CYP-S1; EC 5.2.1.8; epididymis secretory protein Li 39; HEL-S-39; OI9; Peptidyl-prolyl cis-trans isomerase B; peptidylprolyl isomerase B; peptidylprolyl isomerase B (cyclophilin B); PPIase; PPIase B; Ppib; Rotamase; Rotamase B; S-cyclophilin; S-cyclophylin; SCYLP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 19035, 478337, 5479, 64367 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 | |
| P23284, P24368, P24369 | |
| PPIB | |
| Synthetic peptide corresponding to residues C(194) G K I E V E K P F A I A K E(208) of human CyPB. | |
| 50 μL | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.