missing translation for 'onlineSavingsMsg'
Learn More

Cyclophilin B Antibody [PE], Novus Biologicals™

Product Code. 30731873 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30731873 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731873 Supplier Novus Biologicals Supplier No. AF5410P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Cyclophilin B Polyclonal antibody specifically detects Cyclophilin B in Human,Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry,Gene Knock-Out
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclophilin B
Applications Western Blot, Immunocytochemistry, Gene Knock-Out
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias Cyclophilin B, cyclophilin-like protein, CYPBEC 5.2.1.8, CYP-S1, MGC2224, OI9MGC14109, peptidyl-prolyl cis-trans isomerase B, peptidylprolyl isomerase B (cyclophilin B), PPIase B, Rotamase B, S-cyclophilin, SCYLP
Host Species Goat
Immunogen E. coli-derived recombinant human Cyclophilin B, Asp34-Glu216, Accession # P23284
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Loading Controls
Primary or Secondary Primary
Gene ID (Entrez) 5479
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.