missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclophilin B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
£289.00 - £460.00
Specifications
| Antigen | Cyclophilin B |
|---|---|
| Concentration | 0.1 mg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18461672
|
Novus Biologicals
NBP1-85358-25ul |
25 μL |
£289.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18746433
|
Novus Biologicals
NBP1-85358 |
£460.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Cyclophilin B Polyclonal specifically detects Cyclophilin B in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| Cyclophilin B | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| Cyclophilin B, cyclophilin-like protein, CYPBEC 5.2.1.8, CYP-S1, MGC2224, OI9MGC14109, peptidyl-prolyl cis-trans isomerase B, peptidylprolyl isomerase B (cyclophilin B), PPIase B, Rotamase B, S-cyclophilin, SCYLP | |
| PPIB | |
| IgG | |
| Affinity Purified | |
| Specificity of human Cyclophilin B antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| 0.1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Loading Controls | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 5479 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LKHYGPGWVSMANAGKDTNGSQFFITTVKTAWLDGKHVVFGKVLEGMEVVRKVESTKTDSRDKPLKDVIIADCGKIEVEKPFAIAKE | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title