missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclophilin B Antibody (CL3915) [Alexa Fluor™ 700], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59782AF700
This item is not returnable.
View return policy
Description
Cyclophilin B Monoclonal antibody specifically detects Cyclophilin B in Human,Mouse,Rat samples. It is validated for Western Blot,KnockDown
Specifications
| Cyclophilin B | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Cyclophilin B, cyclophilin-like protein, CYPBEC 5.2.1.8, CYP-S1, MGC2224, OI9MGC14109, peptidyl-prolyl cis-trans isomerase B, peptidylprolyl isomerase B (cyclophilin B), PPIase B, Rotamase B, S-cyclophilin, SCYLP | |
| This antibody was developed using a synthetic peptide derived from P23284, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Loading Controls | |
| 5479 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Western Blot, KnockDown | |
| CL3915 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction