Learn More
Invitrogen™ Cyclophilin A Recombinant Rabbit Monoclonal Antibody (24GB905)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557557
 View more versions of this product
Product Code. 30276067
  • £172.00 / 20µL
Description
Cyclophilin A Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Immunophilins are a family of soluble cytosolic receptors capable of binding to one of two major immunosuppressant agents - cyclosporin A (CsA) or FK506. Proteins that bind FK506 are termed FK506 Binding Proteins (FKBPs) and those that bind cyclosporin A are called cyclophilins (CyP). Both CyP:CsA and FKBP:FK506 complexes have been shown to inhibit calcineurin, a calcium and calmodulin dependent protein phosphatase which has been implicated as an important signaling enzyme in T-cell activation, providing a possible mechanism of immunosuppression by CsA and FK506. Immunophilins function as peptidyl prolyl cis-trans-isomerases (PPIase) whose activity is inhibited by their respective immunosuppressant compounds. As PPIase's, immunophilins accelerate folding of some proteins both in vivo and in vitro by catalyzing slow steps in the initial folding and rearrangement of proline containing proteins. Within the cyclophilin family, there are several different proteins which show a high degree of homology including CyPA, CyPB and CyPC. CyPA, also termed CyP-18, is the most abundant and ubiquitous cyclophilin found in all vertebrate tissues and is present in T-cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cyclophilin A
Recombinant Monoclonal
Unconjugated
Ppia
2700098C05; Cphn; CYCA; cyclophilin; Cyclophilin A; Cyclosporin A-binding protein; CyP A; CyP-18; CYPA; CyP-A; CYPH; EC 5.2.1.8; epididymis secretory sperm binding protein Li 69p; HEL-S-69p; HGNC:9253; I79_011869; MGC117158; MGC12404; MGC23397; p1B15; p31; Peptidyl-prolyl cis-trans isomerase A; Peptidyl-prolyl cis-trans isomerase A, N-terminally processed; peptidylprolyl isomerase A; peptidylprolyl isomerase A (cyclophilin A); PPIA; PPIase; PPIase A; rotamase; rotamase A; SP18; T cell cyclophilin
Rabbit
Affinity chromatography
RUO
268373, 5478
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB905
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P17742, P62937
Ppia
A synthesized peptide derived from human Cyclophilin A (aa 100-165).
20 μL
Primary
Human, Mouse