missing translation for 'onlineSavingsMsg'
Learn More

Cyclophilin A Antibody (817815) [mFluor Violet 450 SE], Novus Biologicals™

Product Code. 30733220 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30733220 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733220 Supplier Novus Biologicals Supplier No. FAB3589MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Cyclophilin A Monoclonal antibody specifically detects Cyclophilin A in Human samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclophilin A
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 817815
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias Cyclophilin A, Cyclosporin A-binding protein, CYPAMGC12404, CYPH, EC 5.2.1.8, MGC117158, MGC23397, peptidyl-prolyl cis-trans isomerase A, peptidylprolyl isomerase A (cyclophilin A), PPIase A, Rotamase A, T cell cyclophilin
Host Species Rat
Immunogen E. coli-derived recombinant human Cyclophilin A, Met1-Glu165, Accession # P62937
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5478
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.