missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclophilin A Antibody (001), DyLight 755, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90520IR
This item is not returnable.
View return policy
Description
Cyclophilin A Monoclonal specifically detects Cyclophilin A in Mouse samples. It is validated for ELISA.
Specifications
| Cyclophilin A | |
| Monoclonal | |
| DyLight 755 | |
| Cyclophilin A, Cyclosporin A-binding protein, CYPAMGC12404, CYPH, EC 5.2.1.8, MGC117158, MGC23397, peptidyl-prolyl cis-trans isomerase A, peptidylprolyl isomerase A (cyclophilin A), PPIase A, Rotamase A, T cell cyclophilin | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Cyclophilin A (NP_032933.1; Met 1-Leu 164) . | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Stem Cell Markers | |
| 5478 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 1 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction