missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclophilin 40 Mouse anti-Human, Clone: 1B8, Novus Biologicals™
Mouse Monoclonal Antibody
£244.00 - £361.00
Specifications
| Antigen | Cyclophilin 40 |
|---|---|
| Clone | 1B8 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18254351
|
Novus Biologicals
NBP2-60239-50UL |
50 μL |
£244.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18217113
|
Novus Biologicals
NBP2-60239-100UL |
100 μL |
£361.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclophilin 40 Monoclonal specifically detects Cyclophilin 40 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Cyclophilin 40 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Immunology | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 5481 | |
| Recombinant human PPID (1-370aa) purified from E. coli | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1B8 | |
| Western Blot 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 40 kDa peptidyl-prolyl cis-trans isomerase, 40 kDa peptidyl-prolyl cis-trans isomerase D, cyclophilin D, Cyclophilin-40, Cyclophilin-related protein, CYP40, CYP-40cyclophilin 40, CYPD, EC 5.2.1.8, MGC33096, peptidyl-prolyl cis-trans isomerase D, peptidylprolyl isomerase D, peptidylprolyl isomerase D (cyclophilin D), PPIase D, Rotamase D | |
| PPID | |
| IgG2b κ | |
| Protein A purified |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title