missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentyltriphenylphosphonium bromide, 96%

Product Code. 11477606
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11477606 250 g 250g
11457606 10 g 10g
11467606 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11477606 Supplier Thermo Scientific Alfa Aesar Supplier No. A14647.30
 Restricted Item
Request Bulk or Custom Formats

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 7333-52-0 | C23H24BrP | 411.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7333-52-0
Molecular Formula C23H24BrP
Molecular Weight (g/mol) 411.32
MDL Number MFCD00051880
InChI Key WZYWSVSFFTZZPE-UHFFFAOYSA-M
Synonym cyclopentyltriphenylphosphonium bromide, cyclopentyl triphenyl phosphanium bromide, phosphonium, cyclopentyltriphenyl-, bromide, cyclopentyltriphenylphosphanium bromide, cyclopentyl triphenyl phosphonium bromide, cyclopentyltriphenylphosphoniumbromide, cyclopentyl triphenylphosphonium bromide, phosphonium,cyclopentyltriphenyl-,bromide 1:1, phosphonium,cyclopentyltriphenyl-, bromide 1:1
PubChem CID 2735896
IUPAC Name cyclopentyl(triphenyl)phosphanium;bromide
SMILES [Br-].C1CCC(C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 262°C to 266°C
Quantity 250 g
Beilstein 3580193
Sensitivity Hygroscopic
Formula Weight 411.33
Percent Purity 98%
Chemical Name or Material Cyclopentyltriphenylphosphonium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.