missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopentylmagnesium chloride, 1M in MeTHF, Thermo Scientific™
Click to view available options
Quantity:
100 mL
500 mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Quantity | 500 mL |
UN Number | UN2924 |
Sensitivity | Air and moisture sensitive |
IUPAC Name | magnesium;cyclopentane;chloride |
Formula Weight | 128.88 |
Concentration or Composition (by Analyte or Components) | 1M in MeTHF |
Chemical Name or Material | Cyclopentylmagnesium chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'