missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentylamine, 99+%

Product Code. 10162641 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10162641 100 g Glass bottle 100g
10799181 5 g Glass bottle 5g
10576251 25 g Glass bottle 25g
10125203 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10162641 Supplier Thermo Scientific Acros Supplier No. 111591000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1003-03-8 | C5H11N | 85.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1003-03-8
Molecular Formula C5H11N
Molecular Weight (g/mol) 85.15
MDL Number MFCD00001380
InChI Key NISGSNTVMOOSJQ-UHFFFAOYSA-N
Synonym cyclopentylamine, aminocyclopentane, amino cyclopentane, unii-4259vry3gn, 1cb, cylcopentylamine, cyclopentyl amine, cyclopentyl-amine, n-cyclopentylamine, cyclo-pentyl amine
PubChem CID 2906
IUPAC Name cyclopentanamine
SMILES NC1CCCC1

Specifications

Density 0.8600g/mL
Boiling Point 106°C to 108°C
Flash Point 17°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C5H9NH2
Refractive Index 1.4482 to 1.4502
Quantity 100 g
Beilstein 12,4
Specific Gravity 0.86
Solubility Information Solubility in water: completely miscible with water
Formula Weight 85.15
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Cyclopentylamine, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.