missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentyl methyl ketone, 98%

Product Code. p-7041699
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15442059

Brand: Thermo Scientific Alfa Aesar H64744.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6004-60-0 | C7H12O | 112.17 g/mol

Cyclopentyl methyl ketone acts as a ligand for Suzuki coupling. Further, it reacts with benzo[1,2,5]oxadiazole 1-oxide to prepare 3-methylspiro(chinoxalin-2(3H),1'-cyclopentan)-3-amin-1,4-dioxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentyl methyl ketone acts as a ligand for Suzuki coupling. Further, it reacts with benzo[1,2,5]oxadiazole 1-oxide to prepare 3-methylspiro(chinoxalin-2(3H),1′-cyclopentan)-3-amin-1,4-dioxide.

Solubility
Miscible with water.

Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, reducing agents, acids, nitric acid, hydrogen peroxide and hydrazine.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6004-60-0
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00060799
InChI Key LKENTYLPIUIMFG-UHFFFAOYSA-N
Synonym 1-cyclopentyl-ethanone, cyclopentyl methyl ketone, ethanone, 1-cyclopentyl, acetylcyclopentane, cyclopentylethanone, ketone, cyclopentyl methyl, 1-cyclopentylethan-1-one, ethanone, 1-cyclopentyl-9ci, ethanone,1-cyclopentyl, methyl cyclopentyl ketone
PubChem CID 22326
IUPAC Name 1-cyclopentylethanone
SMILES CC(=O)C1CCCC1

Specifications

Density 0.913
Boiling Point 151°C to 156°C
Refractive Index 1.4435
Quantity 1 g
Beilstein 1853483
Solubility Information Miscible with water.
Formula Weight 112.17
Percent Purity 98%
Chemical Name or Material Cyclopentyl methyl ketone
compliance-icons
Product Identifier
  • Cyclopentyl methyl ketone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Cyclopentyl methyl ketone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.