missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentyl methyl ketone, 98%

Product Code. 15452059
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15452059 5 g 5g
15442059 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15452059 Supplier Thermo Scientific Alfa Aesar Supplier No. H64744.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6004-60-0 | C7H12O | 112.17 g/mol

Cyclopentyl methyl ketone acts as a ligand for Suzuki coupling. Further, it reacts with benzo[1,2,5]oxadiazole 1-oxide to prepare 3-methylspiro(chinoxalin-2(3H),1'-cyclopentan)-3-amin-1,4-dioxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentyl methyl ketone acts as a ligand for Suzuki coupling. Further, it reacts with benzo[1,2,5]oxadiazole 1-oxide to prepare 3-methylspiro(chinoxalin-2(3H),1′-cyclopentan)-3-amin-1,4-dioxide.

Solubility
Miscible with water.

Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, reducing agents, acids, nitric acid, hydrogen peroxide and hydrazine.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6004-60-0
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00060799
InChI Key LKENTYLPIUIMFG-UHFFFAOYSA-N
Synonym 1-cyclopentyl-ethanone, cyclopentyl methyl ketone, ethanone, 1-cyclopentyl, acetylcyclopentane, cyclopentylethanone, ketone, cyclopentyl methyl, 1-cyclopentylethan-1-one, ethanone, 1-cyclopentyl-9ci, ethanone,1-cyclopentyl, methyl cyclopentyl ketone
PubChem CID 22326
IUPAC Name 1-cyclopentylethanone
SMILES CC(=O)C1CCCC1

Specifications

Density 0.913
Boiling Point 151°C to 156°C
Refractive Index 1.4435
Quantity 5 g
Beilstein 1853483
Solubility Information Miscible with water.
Formula Weight 112.17
Percent Purity 98%
Chemical Name or Material Cyclopentyl methyl ketone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.