missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentyl Methyl Ether, TRC

Product Code. 30180277
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180277 10 g 10g
30180121 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30180277 Supplier Toronto Research Chemicals Supplier No. TRCC99233810G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5614-37-9
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
Synonym Ether, cyclopentyl methyl (7CI,8CI), Methoxycyclopentane, CPME, Cyclopentyl methyl ether, Methyl cyclopentyl ether, Methoxycyclopentane, CPME, Methyl Cyclopentyl Ether;
IUPAC Name methoxycyclopentane
SMILES COC1CCCC1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C6H12O/c1-7-6-4-2-3-5-6/h6H,2-5H2,1H3
Formula Weight 100.0888
Chemical Name or Material Cyclopentyl Methyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.