missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentyl methyl ether, stab., 99.9+%

Product Code. 11377099
Change view
Click to view available options
Quantity:
500 mL
1000 mL
2500 mL
Unit Size:
1000mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377099 1000 mL 1000mL
11367099 500 mL 500mL
11387099 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11377099 Supplier Thermo Scientific Alfa Aesar Supplier No. H27349.AU
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5614-37-9 | C6H12O | 100.16 g/mol

Cyclopentyl methyl ether is a hydrophobic ether solvent widely used in Grignard reaction, Friedel-Crafts reaction, Claisen condensation and Beckmann reaction. It plays an important role in extraction and polymerization reactions as well as in surface coatings. It is considered as a replacement for tetrahydrofuran, methyl tert-butyl ether, dioxane and other existing ether solvents due to its excellent properties like high boiling point and relative stability under acidic and basic conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentyl methyl ether is a hydrophobic ether solvent widely used in Grignard reaction, Friedel-Crafts reaction, Claisen condensation and Beckmann reaction. It plays an important role in extraction and polymerization reactions as well as in surface coatings. It is considered as a replacement for tetrahydrofuran, methyl tert-butyl ether, dioxane and other existing ether solvents due to its excellent properties like high boiling point and relative stability under acidic and basic conditions.

Solubility
Miscible with water, alcohols, ketones and hydrocarbons.

Notes
Store in cool place. Air and light sensitive. Incompatible with oxidizing agents, acids, nitric acid, hydrogen peroxide and hydrazine.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5614-37-9
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD08276401
InChI Key SKTCDJAMAYNROS-UHFFFAOYSA-N
Synonym cyclopentyl methyl ether, cyclopentane, methoxy, cpme, cyclopentane,methoxy, unii-4067e5gbkb, cyclopentylmethylether, cylopentylmethylether, methoxycyclopentane, 2-methoxycyclopentyl, cylopentyl-methylether
PubChem CID 138539
IUPAC Name methoxycyclopentane
SMILES COC1CCCC1

Specifications

Melting Point -140°C
Density 0.86
Boiling Point 106°C
Flash Point −1°C (−30°F)
Refractive Index 1.421
Quantity 1000 mL
UN Number UN3271
Solubility Information Miscible with water,alcohols,ketones and hydrocarbons.
Formula Weight 100.16
Percent Purity ≥99.9%
Chemical Name or Material Cyclopentyl methyl ether
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cyclopentyl methyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Cyclopentyl methyl ether

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.