missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentyl isothiocyanate, 98%, Thermo Scientific™

Product Code. 11479283
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479283 5 g 5g
11469283 1 g 1g
11489283 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479283 Supplier Thermo Scientific Alfa Aesar Supplier No. A11745.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It employed as a intermediate for pharmaceutical.

Solubility
Soluble in ethanol, acetone and hydrolyzes with water.

Notes
Moisture Sensitive, store away form water/moisture. Store in cool, dry conditions in well sealed container. Store away from oxidizing agent. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33522-03-1
Molecular Formula C6H9NS
Molecular Weight (g/mol) 127.205
MDL Number MFCD00040877
InChI Key PJOODZCPFADLCI-UHFFFAOYSA-N
Synonym cyclopentyl isothiocyanate, cyclopentane, isothiocyanato, isothiocyanato-cyclopentane, cyclopentanisothiocyanate, cyclopentylisothiocyanate, acmc-20albz
PubChem CID 141792
IUPAC Name isothiocyanatocyclopentane
SMILES C1CCC(C1)N=C=S

Specifications

Density 1.03
Boiling Point 75°C to 76°C (10 mmHg)
Refractive Index 1.523
Quantity 5 g
UN Number UN2922
Beilstein 2715701
Sensitivity Moisture sensitive
Solubility Information Soluble in ethanol,acetone and hydrolyzes with water.
Formula Weight 127.21
Percent Purity 98%
Chemical Name or Material Cyclopentyl isothiocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.