missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentene, 98+%

Product Code. 11334198
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11334198 25 mL 25mL
11324198 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334198 Supplier Thermo Scientific Alfa Aesar Supplier No. 041461.AC
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 142-29-0 | C5H8 | 68.119 g/mol

Cyclopentene is used in organic synthesis for cross-linking resin. It is used as an intermediate in Industries such as Agrochemical, Dyestuff, Pharmaceutical, syntheses material, Chemical. It is used as a monomer for synthesis of plastics and rubber. It is also used for the synthesis of various chemicals, such as 1,1,2-Trimethylcyclohexane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-29-0
Molecular Formula C5H8
Molecular Weight (g/mol) 68.119
MDL Number MFCD00001394
InChI Key LPIQUOYDBNQMRZ-UHFFFAOYSA-N
Synonym 1-cyclopentene, unii-onm2ckv81z, onm2ckv81z, 1-cyclopentene #, cyclopentene, cyclopent-2-en-1-yl, cyclopent-3-en-1-yl, dsstox_cid_9171, wln: l5utj, acmc-209co5
PubChem CID 8882
ChEBI CHEBI:49155
IUPAC Name cyclopentene
SMILES C1CC=CC1

Specifications

Melting Point -135°C
Color Colorless
Density 0.773 g/mL
Boiling Point 44°C to 45°C
Flash Point -29°C (-20°F)
Refractive Index 1.4225
Quantity 25 mL
UN Number UN2246
Beilstein 635707
Sensitivity Air Sensitive
Solubility Information It is soluble in most organic solvents like ethanol and ether. It is immiscible with water.
Formula Weight 68.12
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material Cyclopentene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclopentene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H304-May be fatal if swallowed and enters airways.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P331-Do NOT induce vomiting.
  • P362-Take off contaminated clothing.
Supplemental information
  • MIXTURE LIST-Contain: Cyclopentene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.