missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopentene, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
Description
Cyclopentene is used in the production of cyclopentene oxide, ketamine, cyclopentyl phenol (disinfectant), aldehyde and cyclopentene glycol. It acts as a monomer and is utilized for the synthesis of plastics and in various chemical syntheses. It is also used in the investigation of [2+2] cycloaddition of diamond (001) surfaces with alkene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cyclopentene is used in the production of cyclopentene oxide, ketamine, cyclopentyl phenol (disinfectant), aldehyde and cyclopentene glycol. It acts as a monomer and is utilized for the synthesis of plastics and in various chemical syntheses. It is also used in the investigation of [2+2] cycloaddition of diamond (001) surfaces with alkene.
Solubility
Miscible with ethanol, ether, benzene and organic solvents. Immiscible with water.
Notes
Store in cool place. Air sensitive. Keep container tightly closed in a dry and well-ventilated place.
Cyclopentene is used in the production of cyclopentene oxide, ketamine, cyclopentyl phenol (disinfectant), aldehyde and cyclopentene glycol. It acts as a monomer and is utilized for the synthesis of plastics and in various chemical syntheses. It is also used in the investigation of [2+2] cycloaddition of diamond (001) surfaces with alkene.
Solubility
Miscible with ethanol, ether, benzene and organic solvents. Immiscible with water.
Notes
Store in cool place. Air sensitive. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Melting Point | -135°C |
Density | 0.773 |
Boiling Point | 44°C to 45°C |
Flash Point | −29°C (−20°F) |
Refractive Index | 1.4225 |
Quantity | 100 mL |
UN Number | UN2246 |
Beilstein | 635707 |
Solubility Information | Miscible with ethanol,ether,benzene and organic solvents. Immiscible with water. |
Formula Weight | 68.12 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'