missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentene, 97%

Product Code. 11438863
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438863 500 mL 500mL
11428863 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11438863 Supplier Thermo Scientific Alfa Aesar Supplier No. A11027.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 142-29-0 | C5H8 | 68.119 g/mol

Cyclopentene is used in the production of cyclopentene oxide, ketamine, cyclopentyl phenol (disinfectant), aldehyde and cyclopentene glycol. It acts as a monomer and is utilized for the synthesis of plastics and in various chemical syntheses. It is also used in the investigation of [2+2] cycloaddition of diamond (001) surfaces with alkene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentene is used in the production of cyclopentene oxide, ketamine, cyclopentyl phenol (disinfectant), aldehyde and cyclopentene glycol. It acts as a monomer and is utilized for the synthesis of plastics and in various chemical syntheses. It is also used in the investigation of [2+2] cycloaddition of diamond (001) surfaces with alkene.

Solubility
Miscible with ethanol, ether, benzene and organic solvents. Immiscible with water.

Notes
Store in cool place. Air sensitive. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-29-0
Molecular Formula C5H8
Molecular Weight (g/mol) 68.119
MDL Number MFCD00001394
InChI Key LPIQUOYDBNQMRZ-UHFFFAOYSA-N
Synonym 1-cyclopentene, unii-onm2ckv81z, onm2ckv81z, 1-cyclopentene #, cyclopentene, cyclopent-2-en-1-yl, cyclopent-3-en-1-yl, dsstox_cid_9171, wln: l5utj, acmc-209co5
PubChem CID 8882
ChEBI CHEBI:49155
IUPAC Name cyclopentene
SMILES C1CC=CC1

Specifications

Melting Point -135°C
Density 0.773
Boiling Point 44°C to 45°C
Flash Point −29°C (−20°F)
Refractive Index 1.4225
Quantity 500 mL
UN Number UN2246
Beilstein 635707
Solubility Information Miscible with ethanol,ether,benzene and organic solvents. Immiscible with water.
Formula Weight 68.12
Percent Purity 97%
Chemical Name or Material Cyclopentene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.