missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanone, TRC

Product Code. 30361194
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30361194 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30361194 Supplier Toronto Research Chemicals Supplier No. TRCC98839510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclopentanone
CAS 120-92-3
Molecular Formula C5 H8 O
Quantity 10 g
Synonym 6: PN: WO2007101224 PAGE: 36 claimed sequence, Adipic ketone, Adipin keton, Dumasin, Ketocyclopentane, Ketopentamethylene, NSC 4122
InChI Formula InChI=1S/C5H8O/c6-5-3-1-2-4-5/h1-4H2
SMILES O=C1CCCC1
IUPAC Name cyclopentanone
Molecular Weight (g/mol) 84.12
Formula Weight 84.0575
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.