missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanone, 99+%, pure

Product Code. 10626682 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
250mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10626682 1 L Glass bottle 1 L
10736521 250 mL Glass bottle 250mL
10552524 2.5 L Glass bottle 2.5 L
11994831 10 L Plastic drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10626682 Supplier Thermo Scientific Acros Supplier No. 111530010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 120-92-3 | C5H8O | 84.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-92-3
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00001409
InChI Key BGTOWKSIORTVQH-UHFFFAOYSA-N
Synonym adipic ketone, ketocyclopentane, ketopentamethylene, adipinketon, dumasin, cyclopentanon, oxocyclopentane, cyclopentan-1-one, unii-220w81tn3s, cyclopentyloxy
PubChem CID 8452
ChEBI CHEBI:16486
IUPAC Name cyclopentanone
SMILES O=C1CCCC1

Specifications

Melting Point -51.0°C
Color Colorless to Yellow
Density 0.9500g/mL
Boiling Point 130.0°C to 131.0°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C5H8(=O)
Refractive Index 1.4360 to 1.4380
Quantity 1 L
Beilstein 07,5
Merck Index 15,2738
Specific Gravity 0.95
Solubility Information Solubility in water: 301g/L (20°C). Other solubilities: miscible with alcohol,acetone and ether
Viscosity 0.8 mPa.s (20°C)
Formula Weight 84.12
Percent Purity 99+%
Grade Pure
Physical Form Liquid
Chemical Name or Material Cyclopentanone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cyclopentanone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.