missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanone-2-carbonitrile, TRC

Product Code. 30357827
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357827 100 mg 100mg
30354902 10 mg 10mg
30355239 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30357827 Supplier Toronto Research Chemicals Supplier No. TRCC992075100MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2941-29-9
Molecular Formula C6 H7 N O
Molecular Weight (g/mol) 109.126
Synonym 2-Oxocyclopentanecarbonitrile, 2-Cyano-1-cyclopentanone, 2-Cyanocyclopentanone, 2-Oxocyclopentane-1-carbonitrile, NSC 146861
IUPAC Name 2-oxocyclopentane-1-carbonitrile
SMILES O=C1CCCC1C#N

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C6H7NO/c7-4-5-2-1-3-6(5)8/h5H,1-3H2
Formula Weight 109.053
Chemical Name or Material Cyclopentanone-2-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.