missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanol, 99%

Product Code. 11472298
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472298 100 mL 100mL
11482298 500 mL 500mL
11492298 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11472298 Supplier Thermo Scientific Alfa Aesar Supplier No. A15249.AE
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 96-41-3 | C5H10O | 86.134 g/mol

Cyclopentanol is a versatile compound used as a building block, intermediate and solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentanol is a versatile compound used as a building block, intermediate and solvent.

Solubility
Slightly soluble in water. Also soluble in ether, alcohols and acetone.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-41-3
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
MDL Number MFCD00001363
InChI Key XCIXKGXIYUWCLL-UHFFFAOYSA-N
Synonym hydroxycyclopentane, cyclopentyl alcohol, 1-cyclopentanol, unii-1l43q07tbu, dsstox_cid_13371, dsstox_rid_79066, dsstox_gsid_33371, cydopentanol, 1-hydroxycyclopentyl, 1-cyclopentyl alcohol
PubChem CID 7298
ChEBI CHEBI:16133
IUPAC Name cyclopentanol
SMILES C1CCC(C1)O

Specifications

Melting Point -19°C
Density 0.95
Boiling Point 139°C to 141°C
Flash Point 51°C (123°F)
Odor Unpleasant
Refractive Index 1.453
Quantity 100 mL
UN Number UN2244
Beilstein 1900556
Merck Index 14,2742
Solubility Information Slightly soluble in water. Also soluble in ether,alcohols and acetone.
Formula Weight 86.13
Percent Purity 99%
Chemical Name or Material Cyclopentanol
Show More Show Less
compliance-icons
Product Identifier
  • Cyclopentanol
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contain: Cyclopentanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.