missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanol, 99%, Thermo Scientific Chemicals

Catalog Number 10696482
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
96-41-3
C5H10O
86.13
MFCD00001363
XCIXKGXIYUWCLL-UHFFFAOYSA-N
hydroxycyclopentane, cyclopentyl alcohol, 1-cyclopentanol, unii-1l43q07tbu, dsstox_cid_13371, dsstox_rid_79066, dsstox_gsid_33371, cydopentanol, 1-hydroxycyclopentyl, 1-cyclopentyl alcohol
7298
CHEBI:16133
cyclopentanol
C1CCC(C1)O
This item is not returnable. View return policy
96-41-3
C5H10O
86.13
MFCD00001363
XCIXKGXIYUWCLL-UHFFFAOYSA-N
hydroxycyclopentane, cyclopentyl alcohol, 1-cyclopentanol, unii-1l43q07tbu, dsstox_cid_13371, dsstox_rid_79066, dsstox_gsid_33371, cydopentanol, 1-hydroxycyclopentyl, 1-cyclopentyl alcohol
7298
CHEBI:16133
cyclopentanol
C1CCC(C1)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -19°C
Color Colorless to Yellow
Density 0.9400g/mL
Boiling Point 139°C to 140°C
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C5H9OH
Refractive Index 1.4520 to 1.454
Quantity 500 g
Beilstein 06, 5
Merck Index 15, 2737
Specific Gravity 0.94
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol
Formula Weight 86.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Cyclopentanol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Cyclopentanol
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'