missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopentanethiol, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Cyclopentanethiol has used as a food spice. It is a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cyclopentanethiol has used as a food spice. It is a pharmaceutical intermediate.
Solubility
Slightly soluble in water; soluble in alcohol and oils.
Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
Cyclopentanethiol has used as a food spice. It is a pharmaceutical intermediate.
Solubility
Slightly soluble in water; soluble in alcohol and oils.
Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
Specifications
Density | 0.957 |
Boiling Point | 131°C to 132°C |
Flash Point | 25°C (77°F) |
Odor | Stench |
Refractive Index | 1.49 |
Quantity | 5 g |
UN Number | UN3336 |
Beilstein | 2343861 |
Sensitivity | Air Sensitive |
Solubility Information | Slightly soluble in water; soluble in alcohol and oils. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'