missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanethiol, 97%

Product Code. 11342226
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11342226 25 g 25g
11332226 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342226 Supplier Thermo Scientific Alfa Aesar Supplier No. L03577.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1679-07-8 | C5H10S | 102.20 g/mol

Cyclopentanethiol has used as a food spice. It is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentanethiol has used as a food spice. It is a pharmaceutical intermediate.

Solubility
Slightly soluble in water; soluble in alcohol and oils.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1679-07-8
Molecular Formula C5H10S
Molecular Weight (g/mol) 102.20
MDL Number MFCD00001369
InChI Key WVDYBOADDMMFIY-UHFFFAOYSA-N
Synonym cyclopentyl mercaptan, mercaptocyclopentane, cyclopentylthiol, fema no. 3262, cyclopentyl thiol, cyclopen-tanethiol, cyclopentane thiol, cyclopentylmercaptan, acmc-209dxb, cyclopentanethiol
PubChem CID 15510
IUPAC Name cyclopentanethiol
SMILES SC1CCCC1

Specifications

Density 0.957
Boiling Point 131°C to 132°C
Flash Point 25°C (77°F)
Odor Stench
Refractive Index 1.49
Quantity 25 g
UN Number UN3336
Beilstein 2343861
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water; soluble in alcohol and oils.
Formula Weight 102.2
Percent Purity 97%
Chemical Name or Material Cyclopentanethiol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.