missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanemethanol, 98%

Product Code. 11424673
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424673 5 g 5g
11434673 25 g 25g
11444673 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11424673 Supplier Thermo Scientific Alfa Aesar Supplier No. A11595.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3637-61-4 | C6H12O | 100.161 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3637-61-4
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00001384
InChI Key ISQVBYGGNVVVHB-UHFFFAOYSA-N
Synonym cyclopentanemethanol, cyclopentyl-methanol, cyclopentylcarbinol, cyclopentylmethyl alcohol, cyclopentanmethanol, cylcopentanemethanol, cyclopentan-methanol, cyclopentyl methanol, cyclopentyl carbinol, cyclo-pentanemethanol
PubChem CID 77195
IUPAC Name cyclopentylmethanol
SMILES C1CCC(C1)CO

Specifications

Density 0.925
Boiling Point 162°C to 163°C
Flash Point 62°C (143°F)
Refractive Index 1.458
Quantity 5 g
Beilstein 1919000
Formula Weight 100.16
Percent Purity 98%
Chemical Name or Material Cyclopentanemethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.