missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanemethanol, 98%, Thermo Scientific™

Product Code. 10775021
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 3637-61-4
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00001384
InChI Key ISQVBYGGNVVVHB-UHFFFAOYSA-N
Synonym cyclopentanemethanol, cyclopentyl-methanol, cyclopentylcarbinol, cyclopentylmethyl alcohol, cyclopentanmethanol, cylcopentanemethanol, cyclopentan-methanol, cyclopentyl methanol, cyclopentyl carbinol, cyclo-pentanemethanol
PubChem CID 77195
IUPAC Name cyclopentylmethanol
SMILES C1CCC(C1)CO

Specifications

Color Colorless
Density 0.9200g/mL
Boiling Point 162°C to 163°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C5H9CH2OH
Refractive Index 1.4565 to 1.4585
Quantity 5 g
Beilstein 06, 9
Specific Gravity 0.92
Formula Weight 100.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Cyclopentanemethanol, 96%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'