missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanemethanol, 98%, Thermo Scientific™

Product Code. 10244810 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10244810 10 g Glass bottle 10g
10775021 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10244810 Supplier Thermo Scientific Acros Supplier No. 147150100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 3637-61-4
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00001384
InChI Key ISQVBYGGNVVVHB-UHFFFAOYSA-N
Synonym cyclopentanemethanol, cyclopentyl-methanol, cyclopentylcarbinol, cyclopentylmethyl alcohol, cyclopentanmethanol, cylcopentanemethanol, cyclopentan-methanol, cyclopentyl methanol, cyclopentyl carbinol, cyclo-pentanemethanol
PubChem CID 77195
IUPAC Name cyclopentylmethanol
SMILES C1CCC(C1)CO

Specifications

Color Colorless
Density 0.9200g/mL
Boiling Point 162°C to 163°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C5H9CH2OH
Refractive Index 1.4565 to 1.4585
Quantity 10 g
Beilstein 06, 9
Specific Gravity 0.92
Formula Weight 100.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Cyclopentanemethanol, 96%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.