missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopentaneethanol, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | Cyclopentaneethanol |
| CAS | 766-00-7 |
| Molecular Formula | C7H14O |
| Quantity | 10 g |
| Synonym | 2-Cyclopentaneethanol, 2-Cyclopentylethanol, 2-Cyclopentylethyl Alcohol |
| InChI Formula | InChI=1S/C7H14O/c8-6-5-7-3-1-2-4-7/h7-8H,1-6H2 |
| SMILES | OCCC1CCCC1 |
| IUPAC Name | 2-cyclopentylethanol |
| Molecular Weight (g/mol) | 114.19 |
| Formula Weight | 114.1045 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction