missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopentanecarboxylic Acid, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Cyclopentanecarboxylic Acid |
| CAS | 3400-45-1 |
| Molecular Formula | C6H10O2 |
| Quantity | 25 g |
| Synonym | Cyclopentanoic Acid, Cyclopentylcarboxylic Acid, NSC 59714 |
| InChI Formula | InChI=1S/C6H10O2/c7-6(8)5-3-1-2-4-5/h5H,1-4H2,(H,7,8) |
| SMILES | OC(=O)C1CCCC1 |
| IUPAC Name | cyclopentanecarboxylic acid |
| Molecular Weight (g/mol) | 114.14 |
| Formula Weight | 114.0681 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction