missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanecarboxylic acid, 99%

Product Code. 11429523
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429523 50 g 50g
11419523 10 g 10g
11439523 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429523 Supplier Thermo Scientific Alfa Aesar Supplier No. A12375.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3400-45-1 | C6H10O2 | 114.144 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3400-45-1
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00001371
InChI Key JBDSSBMEKXHSJF-UHFFFAOYSA-N
Synonym cyclopentane carboxylic acid, cyclopentylcarboxylic acid, cyclopentancarboxylic acid, cyclopentanoic acid, unii-z6691vh94a, cyclopentylmethanoic acid, pubchem14153, carboxycyclopentane, cyclopentyl carboxylic acid, acmc-209i4g
PubChem CID 18840
IUPAC Name cyclopentanecarboxylic acid
SMILES C1CCC(C1)C(=O)O

Specifications

Melting Point -7°C
Density 1.052
Boiling Point 215°C to 216°C
Flash Point 93°C (199°F)
Refractive Index 1.454
Quantity 50 g
Beilstein 2039729
Formula Weight 114.14
Percent Purity 99%
Chemical Name or Material Cyclopentanecarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.