missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanecarboxylic Acid, 98+%

Product Code. 10686482 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10686482 25 g Glass bottle 25g
10375740 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10686482 Supplier Thermo Scientific Acros Supplier No. 111490250
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3400-45-1 | C6H10O2 | 114.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3400-45-1
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00001371
InChI Key JBDSSBMEKXHSJF-UHFFFAOYSA-N
Synonym cyclopentane carboxylic acid, cyclopentylcarboxylic acid, cyclopentancarboxylic acid, cyclopentanoic acid, unii-z6691vh94a, cyclopentylmethanoic acid, pubchem14153, carboxycyclopentane, cyclopentyl carboxylic acid, acmc-209i4g
PubChem CID 18840
IUPAC Name cyclopentanecarboxylic acid
SMILES C1CCC(C1)C(=O)O

Specifications

Melting Point 4.0°C
Color Colorless to Yellow
Density 1.0500g/mL
Boiling Point 216.0°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula C5H9CO2H
Refractive Index 1.4530 to 1.5450
Quantity 25 g
Beilstein 09, 6
Specific Gravity 1.05
Solubility Information Solubility in water: 3g/100mL. Other solubilities: soluble in alcohols, methylene chloride, acetone,, toluene
Formula Weight 114.14
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Cyclopentanecarboxylic acid, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • Cyclopentanecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.