missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanecarboxaldehyde, 97%, stabilized

Product Code. 10356644 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10356644 5 g Glass bottle 5g
10663985 1 g Glass bottle 1g
11964781 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10356644 Supplier Thermo Scientific Acros Supplier No. 425910050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 872-53-7 | C6H10O | 98.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-53-7
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.14
InChI Key VELDYOPRLMJFIK-UHFFFAOYSA-N
Synonym cyclopentanecarboxaldehyde, cyclopentanealdehyde, cyclopentancarbaldehyd, cyclopentylformaldehyde, formylcyclopentane, cyclopentyl aldehyde, 1-formylcyclopentane, cyclopentane-1-carboxaldehyde, cyclopentanecaboxaldehyde, cyclopentanecarboaldehyde
PubChem CID 70106
IUPAC Name cyclopentanecarbaldehyde
SMILES C1CCC(C1)C=O

Specifications

Color Colorless to Yellow
Density 0.9190g/mL
Boiling Point 140.0°C to 141.0°C
Flash Point 28°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C5H9CHO
Refractive Index 1.4420 to 1.4460 (20°C, 589nm)
Quantity 5 g
Specific Gravity 0.919
Stabilizer 0.1 wt% hydroquinone
Formula Weight 98.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Cyclopentanecarboxaldehyde
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cyclopentanecarboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.