missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentane, Reagent Grade, 98%, Honeywell

Product Code. 15615610 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 L
Packaging:
Glass Bottle
Unit Size:
100mL
2 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15615610 2 L Glass Bottle 2 L
15695600 100 mL Glass Bottle 100mL
15625610 500 mL Glass Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15615610 Supplier Honeywell Research Chemicals Supplier No. 4597472L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, 98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 287-92-3
Molecular Formula C5H10
Molecular Weight (g/mol) 70.135
MDL Number MFCD00001356
InChI Key RGSFGYAAUTVSQA-UHFFFAOYSA-N
Synonym pentamethylene, ciclopentano, cyclopentan, zyklopentan, unii-t86pb90rnu, hsdb 62, t86pb90rnu, cyclopentanes, cyclopentyl group, hydrocarbons, cyclic c5 and c6
PubChem CID 9253
ChEBI CHEBI:23492
IUPAC Name cyclopentane
SMILES C1CCCC1

Specifications

CAS Max % 100.0000%
Melting Point -94°C (literature)
Color Colorless
Density 0.751 g/mL at 25 °C(lit.)
Boiling Point 50°C (literature)
Flash Point −20°C
Ignition Point 682°F
Odor Solvent like
Packaging Glass Bottle
Linear Formula C5H10
Quantity 2 L
UN Number UN1146
Beilstein 1900195
Formula Weight 70.13
Vapor Pressure 5.3psi (21°C), 9.9psi (38°C)
Percent Purity >97.5%
Grade Reagent
Chemical Name or Material Cyclopentane, Reagent Grade, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.