missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentane, 95%

Product Code. 11457706
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11457706 500 mL 500mL
11447706 100 mL 100mL
11467706 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11457706 Supplier Thermo Scientific Alfa Aesar Supplier No. A14697.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 287-92-3 | C5H10 | 70.135 g/mol

Cyclopentane is used as green blowing agent and involved in the production of polyurethane insulating foam, which is used in refrigerators, freezers, water heaters, construction panels, insulated pipes and roofs. As a lubricant, it finds applications in computer hard drives and outer space equipment due to its low volatile nature. It is widely useful in the preparation of resin, adhesives and pharmaceutical intermediate. It is an additive in gasoline. Since it is a halogen free compound and has zero-ozone depletion potential, it replaces the conventionally used chloro fluoro carbon (CFC) in refrigeration and thermal insulation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentane is used as green blowing agent and involved in the production of polyurethane insulating foam, which is used in refrigerators, freezers, water heaters, construction panels, insulated pipes and roofs. As a lubricant, it finds applications in computer hard drives and outer space equipment due to its low volatile nature. It is widely useful in the preparation of resin, adhesives and pharmaceutical intermediate. It is an additive in gasoline. Since it is a halogen free compound and has zero-ozone depletion potential, it replaces the conventionally used chloro fluoro carbon (CFC) in refrigeration and thermal insulation.

Solubility
Miscible with ethanol, ether and acetone. Slightly miscible with water.

Notes
Avoid heat, direct sunlight, flames and sparks. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 287-92-3
Molecular Formula C5H10
Molecular Weight (g/mol) 70.135
MDL Number MFCD00001356
InChI Key RGSFGYAAUTVSQA-UHFFFAOYSA-N
Synonym pentamethylene, ciclopentano, cyclopentan, zyklopentan, unii-t86pb90rnu, hsdb 62, t86pb90rnu, cyclopentanes, cyclopentyl group, hydrocarbons, cyclic c5 and c6
PubChem CID 9253
ChEBI CHEBI:23492
IUPAC Name cyclopentane
SMILES C1CCCC1

Specifications

Melting Point -95°C
Density 0.748
Boiling Point 47°C to 49°C
Flash Point −37°C (−34°F)
Assay Percent Range 95%
Odor Petrol-like
Refractive Index 1.406
Quantity 500 mL
UN Number UN1146
Beilstein 1900195
Merck Index 14,2741
Solubility Information Miscible with ethanol,ether and acetone. Slightly miscible with water.
Formula Weight 70.14
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Cyclopentane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclopentane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H336-May cause drowsiness or dizziness.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contain: Cyclopentane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.