missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentadienylvanadium tetracarbonyl, 97+%

Product Code. 11314929
Change view
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.5g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11314929 0.5 g 0.5g
11304929 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11314929 Supplier Thermo Scientific Alfa Aesar Supplier No. 081123.02
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12108-04-2 | C9H5O4V | 228.076 g/mol

Employed as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12108-04-2
Molecular Formula C9H5O4V
Molecular Weight (g/mol) 228.076
MDL Number MFCD00014259
InChI Key LIFGJKDEFQAOFY-UHFFFAOYSA-N
Synonym carbon monoxide; cyclopentane; vanadium, tetracarbonyl .eta.5-cyclopenta-2,4-dien-1-yl vanadium
PubChem CID 498652
IUPAC Name carbon monoxide;cyclopentane;vanadium
SMILES [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[CH]1[CH][CH][CH][CH]1.[V]

Specifications

Melting Point 138°C
Boiling Point 80°C to 100°C (0.1 mmHg)(sublimation)
Quantity 0.5 g
UN Number UN3466
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 228.08
Percent Purity ≥97%
Physical Form Crystalline
Chemical Name or Material Cyclopentadienylvanadium tetracarbonyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.