missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentadienylmolybdenum tricarbonyl dimer, 98%

Product Code. 11363638
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11363638 5 g 5g
11383638 1 g 1g
11373638 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11363638 Supplier Thermo Scientific Alfa Aesar Supplier No. 039245.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 12091-64-4 | C16H2Mo2O6 | 482.09 g/mol

It is widely used as a chemical intermediate in the laboratories, manufacture purposes, chemical and pharmaceutical intemediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12091-64-4
Molecular Formula C16H2Mo2O6
Molecular Weight (g/mol) 482.09
MDL Number MFCD00009947
InChI Key AYBJYGWNVNZERR-UHFFFAOYSA-N
Synonym Bis(tricarbonylcyclopentadienylmolybdenum); Tricarbonylcyclopentadienylmolybdenum dimer

Specifications

Melting Point 220°C (decomposition)
Color Purple to Red
Quantity 5 g
UN Number UN3466
Sensitivity Air sensitive
Solubility Information It is insoluble in water.
Formula Weight 490.13
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Cyclopentadienylmolybdenum tricarbonyl dimer

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.