missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentadienylmanganese tricarbonyl

Product Code. 11338927
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338927 5 g 5g
11348927 1 g 1g
11328927 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11338927 Supplier Thermo Scientific Alfa Aesar Supplier No. 038606.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 12079-65-1

Cyclopentadienylmanganese tricarbonyl is an important raw material and intermediate used in organic synthesis, pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12079-65-1
MDL Number MFCD00001440
Synonym Tricarbonylcyclopentadienylmanganese

Specifications

Melting Point 77°C
Physical Form Crystalline
Quantity 5 g
UN Number UN3466
Solubility Information Soluble in water (70 ppm (25C)
Formula Weight 204.06
Sensitivity Air sensitive
Chemical Name or Material Cyclopentadienylmanganese tricarbonyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.