missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentadecane, 98%

Product Code. 11434017
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434017 5 g 5g
11424017 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11434017 Supplier Thermo Scientific Alfa Aesar Supplier No. A19416.06
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 295-48-7 | C15H30 | 210.405 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 295-48-7
Molecular Formula C15H30
Molecular Weight (g/mol) 210.405
MDL Number MFCD00039424
InChI Key SRONXYPFSAKOGH-UHFFFAOYSA-N
Synonym acmc-209h9p
PubChem CID 67525
IUPAC Name cyclopentadecane
SMILES C1CCCCCCCCCCCCCC1

Specifications

Density 0.805
Quantity 5 g
Formula Weight 210.4
Percent Purity 98%
Chemical Name or Material Cyclopentadecane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.