missing translation for 'onlineSavingsMsg'
Learn More

Cyclopamine, TRC

Product Code. 18091250
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18091250 10 mg 10mg
18034050 1 mg 1mg
18013840 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18091250 Supplier Toronto Research Chemicals Supplier No. TRCC98840010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4449-51-8
Molecular Formula C27 H41 N O2
Molecular Weight (g/mol) 411.62
Synonym Spiro[9H-benzo[a]fluorene-9,2'(3'H)-furo[3,2-b]pyridin]-3-ol, 1,2,3,3'a,4,4',5',6,6',6a,6b,7,7',7'a,8,11,11a,11b-octadecahydro-3',6',10,11b-tetramethyl-, (2'R,3S,3'R,3'aS,6'S,6aS,6bS,7'aR,11aS,11bR)-, Cyclopamine (8CI), Jervine, 11-deoxo- (7CI), Veratraman-3-ol, 17,23-epoxy-, (3β,23β)-, (-)-Cyclopamine, 11-Deoxojervine, 11-Deoxyjervine
IUPAC Name (3S,3'R,3'aS,6'S,6aS,6bS,7'aR,9R,11aS,11bR)-3',6',10,11b-tetramethylspiro[2,3,4,6,6a,6b,7,8,11,11a-decahydro-1H-benzo[a]fluorene-9,2'-3a,4,5,6,7,7a-hexahydro-3H-furo[3,2-b]pyridine]-3-ol
SMILES C[C@@H]1CN[C@@H]2[C@@H](C1)O[C@]3(CC[C@H]4[C@@H]5CC=C6C[C@@H](O)CC[C@]6(C)[C@H]5CC4=C3C)[C@@H]2C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C27H41NO2/c1-15-11-24-25(28-14-15)17(3)27(30-24)10-8-20-21-6-5-18-12-19(29)7-9-26(18,4)23(21)13-22(20)16(27)2/h5,15,17,19-21,23-25,28-29H,6-14H2,1-4H3/t15-,17+,19-,20-,21-,23-,24+,25-,26-,27-/m0/s1
Formula Weight 411.3137
Chemical Name or Material Cyclopamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.